CAS RN 1797114-57-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
alpha-[[(1,1-Dimethylethyl)(phenylmethyl)amino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol (N-Benzyl Salbutamol Acetonide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Benzyl Salbutamol Acetonide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Benzyl Salbutamol Acetonide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.