CAS RN 179688-29-0
IUPAC Navn:
6,7-bis(2-methoxyethoxy)-1,4-dihydroquinazolin-4-one
Synonymer:
6,7-bis(2-methoxyethoxy)-1H-quinazolin-4-one
Molekylvægt (g/mol):
294.31
Molekylformel:
C14H18N2O5
InChi Key:
PMQWTUWLIGJTQD-UHFFFAOYSA-N
SMILES:
COCCOC1=C(OCCOC)C=C2C(=O)N=CNC2=C1
1
–
3
af
3
Resultater
1
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6,7-bis(2-Methoxyethoxy)-3,4-dihydroquinazolin-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6,7-bis(2-Methoxyethoxy)-3,4-dihydroquinazolin-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.