Få mere at vide
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one, Mikromol™
Artikelnummer. 30051620
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051620 25 mg
17900666 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051620 Leverandør Mikromol Leverandørnr. MM3297.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6,7-bis(2-Methoxyethoxy)-3,4-dihydroquinazolin-4-one
CAS 179688-29-0
Analyt- eller komponentnavne 6,7-bis(2-Methoxyethoxy)-3,4-dihydroquinazolin-4-one
Forsendelsestilstand Room Temperature
Molekylær formel C14 H18 N2 O5
Certificeringer/compliance ISO 17025
Synonym 6,7-bis(2-methoxyethoxy)-4(3H)-Quinazolinone, 4(1H)-Quinazolinone, 6,7-bis(2-methoxyethoxy)- (9CI), 6,7-Bis(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one, 6,7-Bis(2-methoxyethoxy)-4(3H)-quinazolinone, 6,7-Bis(2-methoxyethoxy)quinazolin-4(3H)-one, 6,7-Di(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one
InChI formel InChI=1S/C14H18N2O5/c1-18-3-5-20-12-7-10-11(15-9-16-14(10)17)8-13(12)21-6-4-19-2/h7-9H,3-6H2,1-2H3,(H,15,16,17)
SMIL COCCOc1cc2N=CNC(=O)c2cc1OCCOC
IUPAC navn 6,7-bis(2-methoxyethoxy)-3H-quinazolin-4-one
Molekylvægt (g/mol) 294.3
Mængde 25 mg
Formel vægt 294.1216 g/mol