CAS RN 1795136-77-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Monocrotophos-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Monocrotophos-d6 (Major), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Monocrotophos-d6 (Major), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Monocrotophos-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.