Få mere at vide
Monocrotophos-d6 (Major), TRC
Artikelnummer. 30473704
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30473704 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473704 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5260532.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Monocrotophos-d6
CAS 1795136-77-4
Renhedsgrad noter HPLC
Molekylær formel C7H8D6NO5P
Mængde 2.5 mg
Synonym Phosphoric Acid Dimethyl-d6 (1E)-1-Methyl-3-(methylamino)-3-oxo-1-propenyl Ester, 3-Hydroxy-N-methyl-cis-crotonamide Dimethyl-d6 Phosphate, Apadrin-d6, Azodrin-d6, Azodrin 202R-d6, Azodrin 60-d6, C 1414-d6, Corophos-d6, Croton 36-d6, Kadett 36-d6, Monicil-d6, Monocil 40-d6, Monocos-d6, Monocron-d6, Monodhan-d6, Monokrotofos-d6, Monostar-d6, Nuvacron-d6, Nuvacron 36SL-d6, Parryfos-d6, SD 9129-d6, Sufos-d6
InChI formel InChI=1S/C7H14NO5P/c1-6(5-7(9)8-2)13-14(10,11-3)12-4/h5H,1-4H3,(H,8,9)/i3D3,4D3
SMIL [2H]C([2H])([2H])OP(=O)(OC(=CC(=O)NC)C)OC([2H])([2H])[2H]
IUPAC navn [1-methyl-3-(methylamino)-3-oxo-prop-1-enyl] bis(trideuteriomethyl) phosphate
Molekylvægt (g/mol) 229.2
Procent renhed >95