CAS RN 1794783-33-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1-[(2'-Cyano-1,1'-biphenyl-4-yl)methyl]-2-ethoxy-d5-7-benzimidazolecarboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[(2'-Cyano-1,1'-biphenyl-4-yl)methyl]-2-ethoxy-d5-7-benzimidazolecarboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.