Få mere at vide
1-[(2'-Cyano-1,1'-biphenyl-4-yl)methyl]-2-ethoxy-d5-7-benzimidazolecarboxylic Acid Methyl Ester, TRC
Artikelnummer. 30366694
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30366694 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366694 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98833250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[(2'-Cyano-1,1'-biphenyl-4-yl)methyl]-2-ethoxy-d5-7-benzimidazolecarboxylic Acid Methyl Ester
CAS 1794783-33-7
Molekylær formel C25H16D5N3O3
Mængde 50 mg
Synonym 3-[(2'-Cyanobiphenyl-4-yl)methyl]-2-ethoxy-d5-3H-benzimidazole-4-carboxylic Acid Methyl Ester, Methyl 1-[(2'-Cyanobiphenyl-4-yl)methyl]-2-ethoxybenzimidazole-7-carboxylate-d5
InChI formel InChI=1S/C25H21N3O3/c1-3-31-25-27-22-10-6-9-21(24(29)30-2)23(22)28(25)16-17-11-13-18(14-12-17)20-8-5-4-7-19(20)15-26/h4-14H,3,16H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C#N
IUPAC navn methyl 3-[[4-(2-cyanophenyl)phenyl]methyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzimidazole-4-carboxylate
Molekylvægt (g/mol) 416.48
Formel vægt 416.1897