CAS RN 177748-19-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(E)-1-[4-[2-(N,N-Dimethylamino)ethoxy]phenyl]-1-[4-(trimethylacetoxy)phenyl]-2-phenylbut-1-ene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(E)-1-[4-[2-(N,N-Dimethylamino)ethoxy]phenyl]-1-[4-(trimethylacetoxy)phenyl]-2-phenylbut-1-ene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(E)-1-[4-[2-(N,N-Dimethylamino)ethoxy]phenyl]-1-[4-(trimethylacetoxy)phenyl]-2-phenylbut-1-ene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.