CAS RN 176258-88-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetamide (3,3',5,5'-Tetraiodothyroacetamide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3,5,3',5'-Tetraiodo Thyroacetamide, TRC
CAS: 176258-88-1 Molekylær formel: C14 H9 I4 N O3 Molekylvægt (g/mol): 746.84 Synonym: Benzeneacetamide, 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo- IUPAC navn: 2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetamide SMIL: NC(=O)Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1