CAS RN 17439-11-1
IUPAC Navn:
titanium(4+) dihydrogen hexafluoride
Synonymer:
hexafluorotitanic acid
Molekylvægt (g/mol):
163.87
Molekylformel:
F6H2Ti
InChi Key:
PFSXARRIPPWGNC-UHFFFAOYSA-J
SMILES:
[H+].[H+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
1
–
1
af
1
Resultater
1
Dihydrogenhexafluortitanat, 60% w/w aq. soln., Thermo Scientific Chemicals
CAS: 17439-11-1 MDL nummer: MFCD00049647 Synonym: Fluorotitanic acid; Hydrogen hexafluorotitanate