CAS RN 17391-18-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Corynoxine B, MedChemExpress
MedChemExpress Corynoxine B is an oxindole alkaloid isolated from Uncaria rhynchophylla (Miq.) Jacks (Gouteng in Chinese); a Beclin-1-dependent autophagy inducer.