CAS RN 17302-01-1
IUPAC Navn:
3-ethyl-3-methylheptane
Synonymer:
3-ethyl-3-methylheptane
Molekylvægt (g/mol):
142.29
Molekylformel:
C10H22
InChi Key:
HSOMNBKXPGCNBH-UHFFFAOYSA-N
SMILES:
CCCCC(C)(CC)CC
1
–
3
af
3
Resultater
1
3-Ethyl-3-methylheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Ethyl-3-methylheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Ethyl-3-methylheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.