CAS RN 172838-30-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Benzyl 3-Amino-3-deoxy-Alpha-D-mannopyranoside Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl 3-Amino-3-deoxy-Alpha-D-mannopyranoside Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzyl 3-Amino-3-deoxy-Alpha-D-mannopyranoside Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.