CAS RN 17135-78-3
IUPAC Navn:
2-chloroanthracene
Synonymer:
2-chloroanthracene
Molekylvægt (g/mol):
212.68
Molekylformel:
C14H9Cl
InChi Key:
OWFINXQLBMJDJQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2C=C3C=CC=CC3=CC2=C1
1
–
3
af
3
Resultater
1
2-Chloroanthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Chloroanthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chloroanthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.