CAS RN 1709-70-2
IUPAC Navn:
4-({3,5-bis[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]-2,4,6-trimethylphenyl}methyl)-2,6-di-tert-butylphenol
Synonymer:
ethanox 330 tris-bht mesitylene
Molekylvægt (g/mol):
775.22
Molekylformel:
C54H78O3
InChi Key:
VSAWBBYYMBQKIK-UHFFFAOYSA-N
SMILES:
CC1=C(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)C(C)=C(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)C(C)=C1CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
1
–
3
af
3
Resultater
1
1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-2,4,6-trimethylbenzene, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.