CAS RN 170365-25-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
KW-8232 free base, MedChemExpress
MedChemExpress KW-8232 free base, an orally active anti-osteoporotic agent, and can reduces the biosynthesis of PGE2.