CAS RN 16816-67-4
IUPAC Navn:
(2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide
Synonymer:
pantethine pantomin D-pantethine pantetina bis(pantothenamidoethyl) disulfide
Molekylvægt (g/mol):
554.72
Molekylformel:
C22H42N4O8S2
InChi Key:
DJWYOLJPSHDSAL-ROUUACIJSA-N
SMILES:
CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
1
–
3
af
3
Resultater
1
D-Pantethine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
D-Pantethine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pantethine, MedChemExpress
MedChemExpress Pantethine is a dimeric form of pantothenic acid, is an intermediate in the production of Coenzyme A, is available as a dietary supplement, and is used to treat acne and improve the blood-cholesterol profile.