CAS RN 168080-93-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2,2,3,3,9,9,10,10-Octamethyl-6-(2-propen-1-yl)-4,8-dioxa-3,9-disilaundecane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2,3,3,9,9,10,10-Octamethyl-6-(2-propen-1-yl)-4,8-dioxa-3,9-disilaundecane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2,3,3,9,9,10,10-Octamethyl-6-(2-propen-1-yl)-4,8-dioxa-3,9-disilaundecane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.