CAS RN 1679333-73-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MMV008138, MedChemExpress
MedChemExpress MMV008138 is a species-selective IspD (enzyme 2-C-methyl-d-erythritol 4-phosphate cytidylyltransferase)-targeting antimalarial agent, with an IC50 of 44 nM for PfIspD (P. falciparum IspD). MMV008138 inhibits the growth of P. falciparum Dd2 strain with an IC50 of 250 nM.