CAS RN 1672-50-0
IUPAC Navn:
5,6-diamino-1,4-dihydropyrimidin-4-one
Synonymer:
5,6-diamino-1H-pyrimidin-4-one
Molekylvægt (g/mol):
126.12
Molekylformel:
C4H6N4O
InChi Key:
PWRHKLKFADDKHS-UHFFFAOYSA-N
SMILES:
NC1=C(N)C(=O)N=CN1
1
–
3
af
3
Resultater
1
5,6-Diamino-4-hydroxypyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,6-Diamino-4-hydroxypyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,6-Diamino-4-hydroxypyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.