CAS RN 166948-47-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(4S)-3-[(2R)-2-[(2R)-2-(2,4-Difluorophenyl)oxiranyl]-1-oxopropyl]-4-benzyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-3-[(2R)-2-[(2R)-2-(2,4-Difluorophenyl)oxiranyl]-1-oxopropyl]-4-benzyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4S)-3-[(2R)-2-[(2R)-2-(2,4-Difluorophenyl)oxiranyl]-1-oxopropyl]-4-benzyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.