CAS RN 16369-14-5
IUPAC Navn:
(2S)-1-hydroxypentan-2-aminium
Synonymer:
(2S)-1-hydroxypentan-2-aminium
Molekylvægt (g/mol):
104.17
Molekylformel:
C5H14NO
InChi Key:
ULAXUFGARZZKTK-YFKPBYRVSA-O
SMILES:
CCC[C@H]([NH3+])CO
1
–
3
af
3
Resultater
1
DL-2-Amino-1-pentanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
DL-2-Amino-1-pentanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
DL-2-Amino-1-pentanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.