CAS RN 1628838-42-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
RAF709, MedChemExpress
MedChemExpress RAF709 is a potent, selective, and efficacious RAF inhibitor with IC50s of 0.4 nM and 0.5 nM for BRAF and CRAF, respectively. Antitumor efficacy.