CAS RN 1624-62-0
IUPAC Navn:
(3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
Synonymer:
(3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-2H,3H,3aH,3bH,4H,5H,9bH,10H,11H-cyclopenta[a]phenanthren-1-one
Molekylvægt (g/mol):
284.4
Molekylformel:
C19H24O2
InChi Key:
BCWWDWHFBMPLFQ-VXNCWWDNSA-N
SMILES:
COC1=CC=C2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCC2=C1
1
–
2
af
2
Resultater
1
Estrone 3-Methyl Ether, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-O-Methyl Estrone, TRC
CAS: 1624-62-0 Molekylær formel: C19 H24 O2 Molekylvægt (g/mol): 284.39 Synonym: Estrone 3-Methyl Ether IUPAC navn: (8R,9S,13S,14S)-3-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one SMIL: COc1ccc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2c1