CAS RN 1620574-24-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
DLK-IN-1, MedChemExpress
MedChemExpress DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.