CAS RN 1620323-43-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
17-(3-Pyridinyl)-3-propanoate-androsta-5,16-dien-3-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
17-(3-Pyridinyl)-3-propanoate-androsta-5,16-dien-3-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
17-(3-Pyridinyl)-3-propanoate-androsta-5,16-dien-3-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.