CAS RN 1616728-72-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(1S,3aR,6aS)-2-[(2S)-2-[[(2S)-2-Cyclohexyl-2-[(2-pyrazinylcarbonyl)amino]acetyl]amino]-3,3-dimethyl-1-oxobutyl]octahydrocyclopenta[c]pyrrole-1-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.