CAS RN 161594-55-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
cis-8-Benzyl-7,9-dioxo-2,8-diazabicyclo[4.3.0]-nonane(Relative stereochemistry), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
cis-8-Benzyl-7,9-dioxo-2,8-diazabicyclo[4.3.0]-nonane(Relative stereochemistry), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-8-Benzyl-7,9-dioxo-2,8-diazabicyclo[4.3.0]-nonane(Relative stereochemistry), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.