CAS RN 1613617-05-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SAHA chloroalkane T1, MedChemExpress
MedChemExpress SAHA chloroalkane T1 is a chloroalkane capture tag by tethering Vorinostat (SAHA) and a chloroalkane tag T1.