CAS RN 1613404-76-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Deltarasin hydrochloride, MedChemExpress
MedChemExpress Deltarasin hydrochloride is an inhibitor of KRAS-PDEδinteraction with Kd of 38 nM for binding to purified PDEδ.