CAS RN 161230-88-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MDL 105519, MedChemExpress
MedChemExpress MDL 105519 is a potent and selective antagonist of glycine binding to the NMDA receptor.