CAS RN 161016-51-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Hosenkoside M, MedChemExpress
MedChemExpress Hosenkoside M is a baccharane glycoside isolated from the seeds of impatiens balsamina.