CAS RN 1609243-64-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
N6-({[3-(Methylsulfinyl)propyl]amino}carbonothioyl)lysine-13C6,15N2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.