CAS RN 16059-16-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2,2,4,4,6,6-Hexakis[(2,2,3,3,4,4,5,5-octafluoropentyl)oxy]-2Lambda5,4Lambda5,6Lambda5-1,3,5,2,4,6-triazatriphosphorine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2,4,4,6,6-Hexakis[(2,2,3,3,4,4,5,5-octafluoropentyl)oxy]-2λ5,4λ5,6λ5-1,3,5,2,4,6-triazatriphosphorine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2,4,4,6,6-Hexakis[(2,2,3,3,4,4,5,5-octafluoropentyl)oxy]-2Lambda5,4Lambda5,6Lambda5-1,3,5,2,4,6-triazatriphosphorine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.