CAS RN 16024-58-1
IUPAC Navn:
2-[2-(2-methoxyethoxy)ethoxy]acetic acid
Synonymer:
[2-(2-methoxyethoxy)ethoxy]acetic acid
Molekylvægt (g/mol):
178.18
Molekylformel:
C7H14O5
InChi Key:
YHBWXWLDOKIVCJ-UHFFFAOYSA-N
SMILES:
COCCOCCOCC(O)=O
1
–
3
af
3
Resultater
1
2-(2-(2-Methoxyethoxy)ethoxy)acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(2-(2-Methoxyethoxy)ethoxy)acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(2-(2-Methoxyethoxy)ethoxy)acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.