CAS RN 16011-96-4
IUPAC Navn:
hydrogen 3,4,5,6-tetrahydropyridin-2-amine chloride
Synonymer:
hydrogen 3,4,5,6-tetrahydropyridin-2-amine chloride
Molekylvægt (g/mol):
134.61
Molekylformel:
C5H11ClN2
InChi Key:
ZHDTXTDHBRADLM-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC1=NCCCC1
1
–
1
af
1
Resultater
1
2-Iminopiperidine, Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.