CAS RN 1598383-41-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
EPZ011989 trifluoroacetate, MedChemExpress
MedChemExpress EPZ011989 trifluoroacetate is a potent, selective orally bioavailable EZH2 inhibitor with Ki 3000-fold selectivity over other HMTase.