CAS RN 159611-02-6
IUPAC Navn:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(furan-2-yl)propanoate
Synonymer:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(furan-2-yl)propanoate
Molekylvægt (g/mol):
376.39
Molekylformel:
C22H18NO5
InChi Key:
AJXDCHXGNUFBRC-FQEVSTJZSA-M
SMILES:
[O-]C(=O)[C@H](CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
af
1
Resultater
1
Fmoc-L-2-furylalanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.