CAS RN 159237-99-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate is an activator of K+ current, with ED50 of 23 nM in the peptidergic caudodorsal neurons.
2
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate is an activator of K+ current, with ED50 of 23 nM in the peptidergic caudodorsal neurons.
3
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate
MedChemExpress Phe-Met-Arg-Phe amide trifluoroacetate is an activator of K+ current, with ED50 of 23 nM in the peptidergic caudodorsal neurons.