CAS RN 1587705-63-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
INH154, MedChemExpress
MedChemExpress INH154 is a highly potent inhibitor for Nek2 and Hec1 binding (INH), with IC50s of 200 nM and 120 nM for INH in Hela and MB468 cells.