Få mere at vide
INH154, MedChemExpress
Artikelnummer. 16495646
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16495646 100 mg
16491117 50 mg
16467617 5 mg
16454868 10 mg
16453266 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16495646 Leverandør Medchem Express Leverandørnr. HY117154
MedChemExpress INH154 is a highly potent inhibitor for Nek2 and Hec1 binding (INH), with IC50s of 200 nM and 120 nM for INH in Hela and MB468 cells.
INH154 is a highly potent inhibitor for Nek2 and Hec1 binding (INH), with IC50s of 200 nM and 120 nM for INH in Hela and MB468 cells.
Tekniske data
Kemisk navn eller materiale INH154
CAS 1587705-63-2
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C22H24N4OS
Mængde 100 mg
Opløselighedsinformation DMSO : 83.33 mg/mL (212.29 mM; Need ultrasonic)
SMIL O=C(C1=CC=NC=C1)NC2=NC(C3=C(C)C=C(N4CCCCC4)C=C3C)=CS2
Molekylvægt (g/mol) 392.52
Formel vægt 392.52
Procent renhed 98.92%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease