CAS RN 1579991-61-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Chitinase-IN-1, MedChemExpress
MedChemExpress Chitinase-IN-1 is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide; 50 uM/20 uM compound concentration's inhibitory percentage are 75%/67% for chitinase/N- acetyl-hexosaminidase respectively.