CAS RN 1544489-04-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
3-((1-Carboxy-2-((5,5-dimethyl-4,5-dihydroisoxazol-3-yl)thio)ethyl)amino)-3-oxopropanoic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-((1-Carboxy-2-((5,5-dimethyl-4,5-dihydroisoxazol-3-yl)thio)ethyl)amino)-3-oxopropanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.