CAS RN 15441-06-2
IUPAC Navn:
methyl 3-[(3-methoxy-3-oxopropyl)disulfanyl]propanoate
Synonymer:
methyl 3-[(3-methoxy-3-oxopropyl)disulfanyl]propanoate
Molekylvægt (g/mol):
238.32
Molekylformel:
C8H14O4S2
InChi Key:
OSZKBWPMEPEYFU-UHFFFAOYSA-N
SMILES:
COC(=O)CCSSCCC(=O)OC
1
–
2
af
2
Resultater
1
Dimethyl 3,3'-Dithiodipropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dimethyl 3,3'-Dithiodipropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.