CAS RN 15414-82-1
IUPAC Navn:
5-ethylimidazolidine-2,4-dione
Synonymer:
5-ethylimidazolidine-2,4-dione
Molekylvægt (g/mol):
128.13
Molekylformel:
C5H8N2O2
InChi Key:
RSBRXBZGVHQUJK-UHFFFAOYNA-N
SMILES:
CCC1NC(=O)NC1=O
1
–
3
af
3
Resultater
1
5-Ethylhydantoin (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Ethylhydantoin (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Ethylhydantoin (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.