CAS RN 1541-67-9
IUPAC Navn:
dodecylidenebis(2-hydroxyethyl)azanium
Synonymer:
N-lauryl diethanolamine
Molekylvægt (g/mol):
272.45
Molekylformel:
C16H34NO2
InChi Key:
ASGZLXKBMOBPBV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC=[N+](CCO)CCO
1
–
3
af
3
Resultater
1
N-Lauryldiethanolamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Lauryldiethanolamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Lauryldiethanolamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.