Få mere at vide
N-Lauryldiethanolamine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Lauryldiethanolamine |
| CAS | 1541-67-9 |
| Molekylær formel | C16 H35 N O2 |
| Mængde | 500 mg |
| Synonym | 2,2'-(Dodecylimino)bisethanol, Ethanol, 2,2'-(dodecylimino)di- (6CI,7CI,8CI), 2,2'-(Dodecylimino)diethanol, 2,2'-(Laurylimino)diethanol, Bis(2-hydroxyethyl)dodecylamine, Bis(2-hydroxyethyl)laurylamine, Bis(hydroxyethyl)dodecylamine, Bis(β-hydroxyethyl)laurylamine, Dodecylbis(2-hydroxyethyl)amine, Dodecylbis(hydroxyethyl)amine, Dodecyldiethanolamine, Dodecyldiethoxylamine, Elest EA, Fentacare 1202, Lauryldiethanolamine, N,N-Bis(2-hydroxyethyl)dodecylamine, N,N-Bis(2-hydroxyethyl)lauramine, N,N-Bis(2-hydroxyethyl)laurylamine, N,N-Bis(hydroxyethyl)dodecanamine, N,N-Bis(hydroxyethyl)laurylamine, N,N-Di(hydroxyethyl)laurylamine, N,N-Dihydroxyethyl-1-Dodecanamine, N-Dodecyldiethanolamine, N-Lauryl-N,N-diethanolamine, NSC 525737, TB 128K |
| InChI formel | InChI=1S/C16H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-17(13-15-18)14-16-19/h18-19H,2-16H2,1H3 |
| SMIL | CCCCCCCCCCCCN(CCO)CCO |
| IUPAC navn | 2-[butyl(2-hydroxyethyl)amino]ethanol |
| Molekylvægt (g/mol) | 273.45 |
| Formel vægt | 273.2668 |