CAS RN 1527-98-6
IUPAC Navn:
(1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
Synonymer:
(1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
Molekylvægt (g/mol):
252.23
Molekylformel:
C10H12N4O4
InChi Key:
IKGRHEWIFBFXPP-IZZDOVSWSA-N
SMILES:
CCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
1
–
6
af
6
Resultater
2
Butyraldehyde 2,4-Dinitrophenylhydrazone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Butyraldehyde 2,4-Dinitrophenylhydrazone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.