CAS RN 152460-08-7
IUPAC Navn:
N''-(2-methyl-5-nitrophenyl)guanidine; nitric acid
Synonymer:
N''-(2-methyl-5-nitrophenyl)guanidine; nitric acid
Molekylvægt (g/mol):
257.21
Molekylformel:
C8H11N5O5
InChi Key:
IINMQQJNRFDBMV-UHFFFAOYSA-N
SMILES:
O[N+]([O-])=O.CC1=C(C=C(C=C1)[N+]([O-])=O)N=C(N)N
1
–
1
af
1
Resultater
1
1-(2-Methyl-5-nitrophenyl)guanidine Nitrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.