CAS RN 15159-65-6
IUPAC Navn:
(2S)-2-azaniumyl-4-bromobutanoate
Synonymer:
(2S)-2-ammonio-4-bromobutanoate
Molekylvægt (g/mol):
182.02
Molekylformel:
C4H8BrNO2
InChi Key:
LMYNKFDVIZBWDF-VKHMYHEASA-N
SMILES:
[NH3+][C@@H](CCBr)C([O-])=O
1
–
1
af
1
Resultater
1
(S)-(+)-2-Amino-4-bromobutyric Acid Hydrobromide (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.